Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmdn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025476.6 | NP_079752.3 | 2 | 301 | ||
GenBank mRNA | AK002442.1 | - | 2 | 184 | ||
AK009059.1 | - | 2 | 158 | |||
AK010421.2 | - | 2 | 178 | |||
AK029446.1 | - | 2 | 161 | |||
AK041619.1 | - | 1 | 1668 | |||
AK160266.1 | - | 2 | 179 | |||
AK213184.1 | - | 1 | 51 | |||
BC025158.1 | - | 2 | 174 | 4023319 | ||
BC096046.1 | - | 2 | 278 | 1397855 | ||
BY275756.1 | - | 2 | 315 |