Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025645.2 | NP_079921.2 | 2 | 237 | ||
GenBank mRNA | AK002371.1 | - | 2 | 201 | ||
AK004277.1 | - | 2 | 167 | |||
AK136135.1 | - | 2 | 167 | |||
AK148443.1 | - | 2 | 176 | |||
AK152287.1 | - | 2 | 201 | |||
AK152352.1 | - | 2 | 195 | |||
AK169605.1 | - | 2 | 183 | |||
BC058365.1 | - | 2 | 169 | 5714866 | ||
BY002298.1 | - | 2 | 237 |