Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025586.3 | NP_079862.1 | 2, 3 | 218 | ||
GenBank mRNA | AK002234.1 | - | 2, 3 | 215 | ||
AK010319.1 | - | 2, 3 | 219 | |||
AK010674.1 | - | 2, 3 | 217 | |||
AK010916.1 | - | 2, 3 | 219 | |||
AK010931.1 | - | 2, 3 | 219 | |||
AK012025.1 | - | 2, 3 | 215 | |||
AK012423.1 | - | 2, 3 | 210 | |||
AK019386.1 | - | 1, 2 | 206 | |||
AK151138.1 | - | 2, 3 | 215 | |||
AK152711.1 | - | 2, 3 | 216 | |||
AK159183.1 | - | 2, 3 | 216 | |||
AK159626.1 | - | 2, 3 | 216 | |||
AK167996.1 | - | 2, 3 | 216 | |||
AK168125.1 | - | 2, 3 | 219 | |||
AK168807.1 | - | 1, 2 | 210 | |||
AK169662.1 | - | 2, 3 | 216 | |||
BC079842.1 | - | 2, 3 | 205 | 5684456 | ||
BC081441.1 | - | 1, 2 | 202 | 6823341 | ||
BC081442.1 | - | 1, 2 | 198 | 6824136 | ||
BC087917.1 | - | 2, 3 | 245 | 30299997 | ||
BC091735.1 | - | 1, 2 | 195 | 3667351 | ||
BC100335.1 | - | 1, 2 | 185 | 30935863 | ||
BC100561.1 | - | 490 | 1361804 |
Gene Symbol: Gm6937 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC089359.1 | - | 556 |