Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025602.3 | NP_079878.1 | 2 | 191 | ||
GenBank mRNA | AK009028.1 | - | 2 | 558 | ||
AK012917.1 | - | 2 | 174 | |||
AK019970.1 | - | 2 | 192 | |||
AK047009.1 | - | 2 | 596 | |||
AK135550.1 | - | 2 | 216 | |||
BC013721.1 | - | 2 | 574 | |||
BC027401.1 | - | 2 | 176 | 4948072 |