Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cypt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025738.2 | NP_080014.2 | 2 | 623 | ||
GenBank mRNA | AF463502.1 | - | 2 | 590 | ||
AK006892.1 | - | 2 | 611 | |||
AK016467.1 | - | 2 | 625 | |||
BC048455.1 | - | 2 | 607 | |||
BC061079.1 | - | 2 | 607 | 6773161 | ||
BU961231.1 | - | 2 | 625 |