Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025725.2 | NP_080001.1 | 1 | 2142 | ||
GenBank mRNA | AK014878.1 | - | 1 | 2140 | ||
AK030023.1 | - | 1 | 2142 | |||
BC125034.1 | - | 1 | 2128 | 40096237 |