Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025800.3 | NP_080076.1 | 5 | 749 | ||
GenBank mRNA | AK002671.1 | - | 5 | 696 | ||
AK004320.1 | - | 4 | 319 | |||
AK015709.1 | - | 526 | ||||
AK028603.1 | - | 5 | 701 | |||
AK032462.1 | - | 5 | 700 | |||
AK053657.1 | - | 5 | 650 | |||
AK077194.1 | - | 5 | 574 | |||
AK150281.1 | - | 5 | 752 | |||
AK207871.1 | - | 4 | 266 | |||
BC069885.1 | - | 5 | 722 | 3671082 | ||
BC069886.1 | - | 5 | 694 | 6310133 |
Gene Symbol: Ppp1r2-ps3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC100445.1 | - | 1 | 573 |