Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026039.3 | NP_080315.1 | 2, 3 | 299 | ||
GenBank mRNA | AK012886.1 | - | 2, 3 | 300 | ||
AK012903.1 | - | 2, 3 | 276 | |||
AK159696.1 | - | 2, 3 | 302 | |||
BC068306.1 | - | 2, 3 | 224 | 30043405 |