Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CBS SIRNA Assays ›
          Gene Symbol:
          CBS CBSL
          Gene Name:
          cystathionine-beta-synthase
          cystathionine-beta-synthase like
          Gene Aliases:
          HIP4
          -
          Chromosome Location:
          Chr.21: 43053190 - 43076868 on Build GRCh38
          Chromosome Location:
          Chr.21: 6444868 - 6468606 on Build GRCh38
          UniGene ID:
          Hs.533013
          Hs.533013
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 7973

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CBS
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000071.2 NP_000062.1 17 2155
          NM_001178008.2 NP_001171479.1 17 2162
          NM_001178009.2 NP_001171480.1 18 1948
          NM_001320298.1 NP_001307227.1 18 2156
          NM_001321072.1 NP_001308001.1 14 2221
          XM_011529773.2 XP_011528075.1 16 2323
          XM_011529774.2 XP_011528076.1 17 2109
          XM_011529775.2 XP_011528077.1 15 2281
          XM_011529776.2 XP_011528078.1 16 2067
          XM_011529777.1 XP_011528079.1 18 2140
          XM_011529781.1 XP_011528083.1 19 2205
          XM_011529782.1 XP_011528084.1 18 2603
          XM_011529783.2 XP_011528085.1 15 2235
          XM_017028489.1 XP_016883978.1 19 2600
          XM_017028490.1 XP_016883979.1 18 2555
          XM_017028491.1 XP_016883980.1 18 1884
          XM_017028492.1 XP_016883981.1 19 1949
          XM_017028493.1 XP_016883982.1 15 2434
          GenBank mRNA AK294608.1 - 19 2027
          AL137314.1 - 14 1682
          BC000440.2 - 17 2025 2820694
          BC010242.1 - 17 2079 4154798
          BC011381.1 - 17 2018 3869615
          BC012319.1 - 6 832
          L00972.1 - 14 2233
          L14577.1 - 17 2068
          L19501.1 - 17 2091
          X82166.1 - 17 2057
          Gene Symbol: CBSL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321073.1 NP_001308002.1 18 1948
          XM_011546094.1 XP_011544396.1 18 2096
          XM_011546095.2 XP_011544397.1 18 2238
          XM_011546097.2 XP_011544399.1 19 2265
          XM_011546098.1 XP_011544400.1 18 2603
          XM_011546099.1 XP_011544401.1 17 2054
          XM_017028209.1 XP_016883698.1 15 2259
          XM_017028210.1 XP_016883699.1 16 2045
          XM_017028211.1 XP_016883700.1 19 2601
          XM_017028212.1 XP_016883701.1 20 2657
          XM_017028213.1 XP_016883702.1 19 1882
          XM_017028214.1 XP_016883703.1 18 2556
          XM_017028215.1 XP_016883704.1 18 2171
          XM_017028216.1 XP_016883705.1 17 2561
          XM_017028217.1 XP_016883706.1 18 1840
          XM_017028218.1 XP_016883707.1 19 2009
          GenBank mRNA BC007257.1 - 18 1918 3028099

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          lyase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          cysteine biosynthetic process from serine
          L-serine metabolic process
          L-serine catabolic process
          cellular amino acid biosynthetic process
          cysteine biosynthetic process via cystathionine
          cysteine biosynthetic process
          transsulfuration
          L-cysteine catabolic process
          DNA protection
          homocysteine catabolic process
          homocysteine metabolic process
          oxidation-reduction process
          hydrogen sulfide biosynthetic process
          cystathionine beta-synthase activity
          cysteine synthase activity
          protein binding
          oxygen binding
          enzyme binding
          heme binding
          pyridoxal phosphate binding
          ubiquitin protein ligase binding
          identical protein binding
          protein homodimerization activity
          metal ion binding
          nitrite reductase (NO-forming) activity
          carbon monoxide binding
          nitric oxide binding
          modified amino acid binding
          S-adenosyl-L-methionine binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0