Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CLN3 SIRNA Assays ›
          Gene Symbol:
          CLN3
          Gene Name:
          CLN3, battenin
          Gene Aliases:
          BTS, JNCL
          Chromosome Location:
          Chr.16: 28466653 - 28492302 on Build GRCh38
          UniGene ID:
          Hs.534667
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 7977

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CLN3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000086.2 NP_000077.1 11, 12 1231
          NM_001042432.1 NP_001035897.1 12, 13 1267
          NM_001286104.1 NP_001273033.1 11, 12 1195
          NM_001286105.1 NP_001273034.1 9, 10 1072
          NM_001286109.1 NP_001273038.1 9, 10 1080
          NM_001286110.1 NP_001273039.1 10, 11 1152
          GenBank mRNA AF015593.1 - 11, 12 962
          AF015594.1 - 10, 11 923
          AF015595.1 - 9, 10 782
          AF015598.1 - 9, 10 716
          AF015600.1 - 7, 8 638
          AF015601.1 - 6, 7 552
          AF077956.1 - 11, 12 908
          AF077957.1 - 11, 12 908
          AF077958.1 - 11, 12 908
          AF077959.1 - 11, 12 908
          AF077960.1 - 10, 11 764
          AF077961.1 - 11, 12 908
          AF077962.1 - 11, 12 908
          AF077963.1 - 11, 12 908
          AF077964.1 - 9, 10 691
          AF077965.1 - 8, 9 578
          AF077966.1 - 11, 12 908
          AF077967.1 - 10, 11 828
          AF077968.1 - 9, 10 691
          AF077969.1 - 7, 8 522
          AF077970.1 - 7, 8 522
          AF077971.1 - 11, 12 908
          AF077973.1 - 9, 10 749
          AF078169.1 - 11, 12 908
          AK294070.1 - 10, 11 1076
          AK297690.1 - 9, 10 1014
          AK302027.1 - 11, 12 935
          AK302138.1 - 11, 12 956
          AK313002.1 - 12, 13 1039
          BC002394.2 - 12, 13 1025 2820893
          BC004433.1 - 12, 13 1025 2820893
          BC111068.1 - 9, 10 1010 5764535
          EF587244.1 - 4, 5 285
          U32680.1 - 11, 12 1045

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane traffic protein membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          action potential
          globoside metabolic process
          cellular amino acid metabolic process
          ceramide metabolic process
          glucosylceramide metabolic process
          galactosylceramide metabolic process
          sphingomyelin metabolic process
          receptor-mediated endocytosis
          vacuolar transport
          lysosome organization
          lysosomal lumen acidification
          associative learning
          arginine transport
          macroautophagy
          negative regulation of macroautophagy
          protein processing
          protein catabolic process
          ionotropic glutamate receptor signaling pathway
          lysosomal lumen pH elevation
          neurotransmitter metabolic process
          amyloid precursor protein catabolic process
          negative regulation of apoptotic process
          negative regulation of catalytic activity
          negative regulation of neuron apoptotic process
          negative regulation of proteolysis
          vesicle transport along microtubule
          neuromuscular process controlling balance
          regulation of cytosolic calcium ion concentration
          membrane organization
          autophagosome maturation
          protein binding
          unfolded protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0