Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other COL1A1 SIRNA Assays ›
          Gene Symbol:
          COL1A1
          Gene Name:
          collagen type I alpha 1
          Gene Aliases:
          EDSC, OI1, OI2, OI3, OI4
          Chromosome Location:
          Chr.17: 50184096 - 50201648 on Build GRCh38
          UniGene ID:
          Hs.172928
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 7978

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: COL1A1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000088.3 NP_000079.2 51 5410
          XM_005257058.3 XP_005257115.2 49 5149
          XM_005257059.4 XP_005257116.2 38 4501
          XM_011524341.1 XP_011522643.1 48 5221
          GenBank mRNA Z74615.1 - 51 5406

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          extracellular matrix structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          blood vessel development
          osteoblast differentiation
          intramembranous ossification
          endochondral ossification
          blood coagulation
          visual perception
          sensory perception of sound
          positive regulation of epithelial to mesenchymal transition
          negative regulation of cell-substrate adhesion
          protein transport
          platelet activation
          extracellular matrix organization
          collagen fibril organization
          positive regulation of cell migration
          collagen catabolic process
          response to corticosteroid
          response to estradiol
          collagen biosynthetic process
          protein localization to nucleus
          tooth mineralization
          response to drug
          response to hydrogen peroxide
          response to peptide hormone
          skin morphogenesis
          cellular response to fibroblast growth factor stimulus
          tooth eruption
          positive regulation of transcription, DNA-templated
          embryonic skeletal system development
          regulation of immune response
          leukocyte migration
          response to cAMP
          response to hyperoxia
          face morphogenesis
          bone trabecula formation
          cartilage development involved in endochondral bone morphogenesis
          protein heterotrimerization
          cellular response to amino acid stimulus
          cellular response to mechanical stimulus
          cellular response to retinoic acid
          cellular response to vitamin E
          cellular response to tumor necrosis factor
          cellular response to epidermal growth factor stimulus
          cellular response to transforming growth factor beta stimulus
          positive regulation of canonical Wnt signaling pathway
          cellular response to fluoride
          extracellular matrix structural constituent
          protein binding
          identical protein binding
          metal ion binding
          platelet-derived growth factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0