Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026187.5 | NP_080463.3 | 8 | 1554 | ||
GenBank mRNA | AK004948.2 | - | 6 | 1984 | ||
AK012672.1 | - | 1 | 127 | |||
AK156255.1 | - | 8 | 1516 | |||
AK157405.1 | - | 8 | 1516 | |||
BC039910.1 | - | 6 | 2064 | |||
BC057896.1 | - | 3 | 396 | |||
BC071240.1 | - | 8 | 1484 | |||
BC150879.1 | - | 8 | 1504 | 9087790 | ||
CX232548.1 | - | 2 | 277 |