Search
Search

Slc25a30| Gene Symbol: Slc25a30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026232.3 | NP_080508.1 | 9 | 978 | ||
| GenBank mRNA | AK015679.1 | - | 9 | 979 | ||
| AK017035.1 | - | 9 | 979 | |||
| AK030950.1 | - | 9 | 947 | |||
| AK090086.1 | - | 9 | 915 | |||
| AK151334.1 | - | 9 | 947 | |||
| AK151420.1 | - | 9 | 915 | |||
| AK159122.1 | - | 9 | 976 | |||
| AK159732.1 | - | 9 | 976 | |||