Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dalrd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026378.3 | NP_080654.3 | 9 | 1260 | ||
GenBank mRNA | AK002439.1 | - | 9 | 1256 | ||
AK003374.1 | - | 3 | 207 | |||
AK149963.1 | - | 9 | 1261 | |||
AK169405.1 | - | 9 | 1258 | |||
AK171818.1 | - | 9 | 1261 | |||
BC013483.1 | - | 9 | 1241 | 4242135 | ||
BC058774.1 | - | 7 | 659 |