siRNA Details
Gene Symbol: Rnf115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026406.3 | NP_080682.3 | 1 | 284 | ||
GenBank mRNA | AK008329.1 | - | 1 | 138 | ||
AK011584.1 | - | 1 | 214 | |||
AK042216.1 | - | 1 | 224 | |||
AK131966.1 | - | 1 | 204 | |||
BC023113.1 | - | 1 | 231 | 4015975 |

More Information
Other Species Targeted

