Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159635.1 | NP_001153107.1 | 2 | 317 | ||
NM_026434.4 | NP_080710.1 | 2 | 317 | |||
GenBank mRNA | AK008110.1 | - | 2 | 173 | ||
AK021243.1 | - | 2 | 198 | |||
AK035348.1 | - | 2 | 225 | |||
AK036220.1 | - | 1 | 2443 | |||
AK036252.1 | - | 2 | 198 | |||
AK135109.1 | - | 2 | 225 | |||
BB841783.1 | - | 2 | 226 | |||
BC005540.1 | - | 2 | 211 | 3590164 | ||
BY009724.1 | - | 2 | 224 | |||
CB183820.1 | - | 2 | 388 |