Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam32a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026455.4 | NP_080731.1 | 2, 3 | 272 | ||
GenBank mRNA | AB041658.1 | - | 2, 3 | 270 | ||
AK007524.1 | - | 2, 3 | 270 | |||
AK008285.1 | - | 2, 3 | 251 | |||
AK008336.1 | - | 2, 3 | 258 | |||
AK011922.1 | - | 2, 3 | 258 | |||
AK075976.1 | - | 2, 3 | 273 | |||
AK082792.1 | - | 2, 3 | 243 | |||
AK150416.1 | - | 2, 3 | 243 | |||
BC006058.1 | - | 2, 3 | 224 | 3591395 |