Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000255.3 | NP_000246.2 | 2 | 350 | ||
XM_005249143.3 | XP_005249200.1 | 2 | 255 | |||
GenBank mRNA | AK292568.1 | - | 2 | 326 | ||
AK312611.1 | - | 2 | 325 | |||
BC016282.1 | - | 2 | 213 | 3908548 | ||
BT007434.1 | - | 1 | 85 | |||
BX647789.1 | - | 2 | 175 | |||
DA777441.1 | - | 2 | 356 | |||
DB290266.1 | - | 2 | 340 | |||
KF030882.1 | - | 1 | 85 | |||
KU178201.1 | - | 1 | 85 | |||
KU178202.1 | - | 1 | 85 | |||
M65131.1 | - | 2 | 161 |