Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026577.3 | NP_080853.3 | 3 | 492 | ||
GenBank mRNA | AK016794.1 | - | 3 | 438 | ||
AK020859.1 | - | 3 | 433 | |||
AK021270.1 | - | 3 | 494 | |||
AK051159.1 | - | 3 | 450 | |||
BC082574.1 | - | 3 | 492 | 30550373 |