Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026613.4 | NP_080889.3 | 4 | 652 | ||
GenBank mRNA | AK011768.1 | - | 3 | 168 | ||
AK015873.1 | - | 3 | 489 | |||
AK033555.1 | - | 4 | 631 | |||
AK140947.1 | - | 1 | 2560 | |||
AK147056.1 | - | 4 | 653 | |||
BC049229.1 | - | 4 | 385 | |||
BC062248.1 | - | 4 | 598 | |||
BC089458.1 | - | 6 | 817 | |||
BC108339.1 | - | 4 | 635 | |||
BC119347.1 | - | 3 | 197 | 8734096 | ||
BC120580.1 | - | 3 | 197 | 40129503 |