Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrriq4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290510.1 | NP_001277439.1 | 2 | 420 | ||
NM_026668.2 | NP_080944.2 | 4 | 583 | |||
GenBank mRNA | AK015024.1 | - | 2 | 371 | ||
AK019775.1 | - | 4 | 584 | |||
BC145806.1 | - | 4 | 284 | 40131109 |