Search
Search

Cep57| Gene Symbol: Cep57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026665.4 | NP_080941.3 | 3 | 475 | ||
| GenBank mRNA | AK014873.1 | - | 3 | 422 | ||
| AK016484.1 | - | 3 | 259 | |||
| AK019414.1 | - | 3 | 355 | |||
| AK028408.1 | - | 3 | 324 | |||
| AK028458.1 | - | 3 | 303 | |||
| AK129053.1 | - | 3 | 424 | |||
| AK167523.1 | - | 3 | 360 | |||
| AY225093.1 | - | 3 | 268 | |||
| AY251192.1 | - | 3 | 420 | |||
| BC025588.1 | - | 2 | 255 | |||
| BC030353.1 | - | 3 | 746 | |||
| BC056968.1 | - | 3 | 721 | |||