Search
Search

Wars2| Gene Symbol: Wars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027462.4 | NP_081738.2 | 2 | 206 | ||
| GenBank mRNA | AK017602.1 | - | 2 | 217 | ||
| AK033774.1 | - | 204 | ||||
| AK036902.1 | - | 2 | 226 | |||
| AK042715.1 | - | 2 | 207 | |||
| AK042964.1 | - | 2 | 207 | |||
| BC089583.1 | - | 2 | 228 | 6762967 | ||