Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142701.1 | NP_001136173.1 | 3 | 648 | ||
NM_027521.3 | NP_081797.1 | 2 | 200 | |||
GenBank mRNA | AK044860.1 | - | 2 | 162 | ||
AK137810.1 | - | 1 | 1420 | |||
AK159194.1 | - | 3 | 649 | |||
AK159474.1 | - | 3 | 581 | |||
AK170262.1 | - | 3 | 634 | |||
AK171304.1 | - | 3 | 634 | |||
BC053750.1 | - | 2 | 200 | 6333971 |