Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027349.3 | NP_081625.3 | 5 | 568 | ||
GenBank mRNA | AK011403.1 | - | 5 | 549 | ||
AK151910.1 | - | 5 | 520 | |||
AK162135.1 | - | 661 | ||||
AK164367.1 | - | 5 | 533 | |||
BC010792.1 | - | 4 | 314 | |||
BC023749.1 | - | 3 | 277 | |||
BC158104.1 | - | 4 | 439 | 9007388 |