Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsll3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027344.3 | NP_081620.2 | 8 | 1106 | ||
GenBank mRNA | AK009935.1 | - | 3 | 501 | ||
AK145382.1 | - | 8 | 1110 | |||
BC147423.1 | - | 7 | 1045 | 9056481 | ||
BC147424.1 | - | 7 | 1045 | 9056499 |