Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rufy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289774.1 | NP_001276703.1 | 5, 6 | 963 | ||
NM_001289775.1 | NP_001276704.1 | 5, 6 | 963 | |||
NM_001289776.1 | NP_001276705.1 | 6, 7 | 1385 | |||
NM_001289777.1 | NP_001276706.1 | 7, 8 | 1210 | |||
NM_027530.3 | NP_081806.1 | 5, 6 | 1331 | |||
GenBank mRNA | AK013185.1 | - | 7, 8 | 1211 | ||
AK018184.1 | - | 5, 6 | 1309 | |||
AK083242.1 | - | 5, 6 | 1266 | |||
AK148795.1 | - | 5, 6 | 627 | |||
AK154941.1 | - | 5, 6 | 964 | |||
AK160820.1 | - | 5, 6 | 964 | |||
BC017648.1 | - | 5, 6 | 720 | 4527858 | ||
BC049127.1 | - | 5, 6 | 1270 | 6821567 | ||
BC058259.1 | - | 6, 7 | 1104 | 6469137 | ||
CT010404.1 | - | 5, 6 | 704 |