Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027563.3 | NP_081839.1 | 7 | 1434 | ||
GenBank mRNA | AK014631.1 | - | 7 | 1436 | ||
BC055948.1 | - | 1 | 58 | |||
BC125420.1 | - | 7 | 1412 | 40129935 | ||
BC125446.1 | - | 7 | 1412 | 40129961 |