Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000303.2 | NP_000294.1 | 2 | 237 | ||
XM_005255374.4 | XP_005255431.1 | 2 | 237 | |||
GenBank mRNA | AK291537.1 | - | 2 | 208 | ||
AK296123.1 | - | 2 | 230 | |||
AK296630.1 | - | 2 | 183 | |||
AK299979.1 | - | 2 | 208 | |||
AK300785.1 | - | 2 | 213 | |||
AK304320.1 | - | 2 | 200 | |||
AK311035.1 | - | 2 | 208 | |||
AK312532.1 | - | 2 | 208 | |||
BC008310.1 | - | 2 | 179 | 3611382 | ||
DC366756.1 | - | 2 | 244 | |||
KU178265.1 | - | 2 | 167 | |||
KU178266.1 | - | 2 | 167 | |||
U85773.1 | - | 2 | 215 |