Search
Search

Chst14| Gene Symbol: Chst14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028117.3 | NP_082393.3 | 1 | 1830 | ||
| GenBank mRNA | AK011230.1 | - | 1 | 1767 | ||
| AK154555.1 | - | 1 | 1819 | |||
| AK155115.1 | - | 2 | 1738 | |||
| BC026886.1 | - | 1 | 1651 | |||
| BC043700.1 | - | 1 | 1803 | 5346204 | ||
| BC085479.1 | - | 1 | 1747 | 6415106 | ||