Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028716.4 | NP_082992.1 | 2 | 161 | ||
GenBank mRNA | AK014030.1 | - | 2 | 210 | ||
AK014380.1 | - | 2 | 162 | |||
BC027776.1 | - | 2 | 156 | 5352251 |