Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166177.1 | NP_001159649.1 | 5 | 1020 | ||
NM_028710.3 | NP_082986.3 | 14 | 2747 | |||
GenBank mRNA | AK018132.2 | - | 11 | 2164 | ||
AK158726.1 | - | 14 | 2748 | |||
AK173082.1 | - | 11 | 1846 | |||
BC022158.1 | - | 10 | 1577 | |||
BC039629.1 | - | 5 | 977 | |||
BC084731.1 | - | 11 | 2437 | 6813261 |