Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293672.1 | NP_001280601.1 | 7 | 1534 | ||
NM_028836.4 | NP_083112.1 | 6 | 1372 | |||
GenBank mRNA | AK008791.2 | - | 6 | 1347 |