Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TNNT2 SIRNA Assays ›
          Gene Symbol:
          TNNT2
          Gene Name:
          troponin T2, cardiac type
          Gene Aliases:
          CMD1D, CMH2, CMPD2, LVNC6, RCM3, TnTC, cTnT
          Chromosome Location:
          Chr.1: 201359008 - 201377828 on Build GRCh38
          UniGene ID:
          Hs.533613
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 8332

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TNNT2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000364.3 NP_000355.2 8, 9 333
          NM_001001430.2 NP_001001430.1 7, 8 303
          NM_001001431.2 NP_001001431.1 7, 8 303
          NM_001001432.2 NP_001001432.1 6, 7 288
          NM_001276345.1 NP_001263274.1 8, 9 333
          NM_001276346.1 NP_001263275.1 8, 9 330
          NM_001276347.1 NP_001263276.1 7, 8 436
          XM_006711508.3 XP_006711571.1 7, 8 333
          XM_006711509.3 XP_006711572.1 7, 8 428
          XM_011509938.2 XP_011508240.1 8, 9 365
          XM_011509939.1 XP_011508241.1 8, 9 377
          XM_011509940.2 XP_011508242.1 8, 9 381
          XM_011509941.2 XP_011508243.1 8, 9 378
          XM_011509942.2 XP_011508244.1 6, 7 335
          XM_011509943.2 XP_011508245.1 6, 7 319
          XM_011509944.2 XP_011508246.1 6, 7 332
          XM_011509946.1 XP_011508248.1 7, 8 406
          XM_017002216.1 XP_016857705.1 7, 8 311
          XM_017002217.1 XP_016857706.1 7, 8 496
          GenBank mRNA AJ709280.1 - 7, 8 323
          AK125236.1 - 7 1450
          AK290621.1 - 7, 8 498
          AK309493.1 - 1, 2 137
          AY277394.1 - 5, 6 246
          BC002653.2 - 7, 8 269 3612968
          L40162.1 - 7, 8 264
          S64668.1 - 6, 7 223
          X74819.1 - 6, 7 261
          X79855.1 - 7, 8 241
          X79856.1 - 7, 8 241
          X79859.1 - 5, 6 465
          X79861.1 - 6, 7 253

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of heart contraction
          muscle filament sliding
          negative regulation of ATPase activity
          positive regulation of ATPase activity
          regulation of muscle filament sliding speed
          protein heterooligomerization
          response to calcium ion
          actin crosslink formation
          ventricular cardiac muscle tissue morphogenesis
          cardiac muscle contraction
          actin binding
          protein binding
          tropomyosin binding
          troponin C binding
          protein binding, bridging
          troponin I binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0