Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syvn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164709.1 | NP_001158181.1 | 16 | 2798 | ||
NM_028769.5 | NP_083045.4 | 16 | 3165 | |||
GenBank mRNA | AK004688.2 | - | 16 | 2652 | ||
AK122558.1 | - | 12 | 3619 | |||
AK143720.1 | - | 16 | 2680 | |||
AK172097.1 | - | 10 | 3795 | |||
BB676375.1 | - | 1 | 114 | |||
BC032931.1 | - | 5 | 1451 | |||
BC042199.1 | - | 16 | 2660 | 4953165 | ||
BC046829.1 | - | 16 | 2798 | 6811574 | ||
BC057917.1 | - | 16 | 3079 | 5136398 | ||
BC080722.1 | - | 16 | 2683 | 5323695 |