Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Robo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF536772.1 | - | 18 | 3418 | ||
AK004723.2 | - | 18 | 3394 | |||
AK030973.1 | - | 2185 | ||||
AK087355.1 | - | 17 | 3570 | |||
AK172104.1 | - | 17 | 3656 | |||
BC071193.1 | - | 17 | 3108 | 5060473 |