Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000405.4 | NP_000396.2 | 2, 3 | 416 | ||
NM_001167607.1 | NP_001161079.1 | 2, 3 | 416 | |||
GenBank mRNA | AK225978.1 | - | 2, 3 | 344 | ||
AK298867.1 | - | 2, 3 | 283 | |||
AK312494.1 | - | 2, 3 | 311 | |||
BC009273.2 | - | 2, 3 | 280 | 4053681 | ||
BX473154.1 | - | 2, 3 | 394 | |||
L01439.1 | - | 2, 3 | 341 | |||
M76477.1 | - | 2, 3 | 341 | |||
X16087.1 | - | 2, 3 | 209 | |||
X61094.1 | - | 1, 2 | 126 | |||
X61095.1 | - | 2, 3 | 272 | |||
X62078.1 | - | 2, 3 | 309 |