siRNA Details
Gene Symbol: 5031410I06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207657.3 | NP_997540.2 | 4 | 541 | ||
GenBank mRNA | AK077239.1 | - | 542 |
Gene Symbol: Gm10220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134299.1 | NP_001127771.1 | 4 | 593 |
Gene Symbol: Gm10471 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177579.1 | NP_001171050.1 | 4 | 559 | ||
GenBank mRNA | BC145097.1 | - | 408 | 9053645 |
Gene Symbol: Gm1979 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281519.1 | NP_001268448.1 | 4 | 518 |
Gene Symbol: Gm21671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281516.1 | NP_001268445.1 | 4 | 518 |
Gene Symbol: Speer4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029376.2 | NP_083652.2 | 4 | 540 | ||
GenBank mRNA | AK006439.1 | - | 4 | 544 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
