Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp6r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164159.1 | NP_001157631.1 | 3 | 501 | ||
NM_028999.1 | NP_083275.1 | 4 | 649 | |||
NM_029456.2 | NP_083732.1 | 4 | 649 | |||
GenBank mRNA | AK015925.1 | - | 4 | 630 | ||
AK018652.1 | - | 3 | 490 | |||
AK035236.1 | - | 4 | 629 | |||
AK088526.1 | - | 3 | 469 | |||
AK129392.1 | - | 4 | 591 | |||
BC008529.1 | - | 4 | 632 | 3601021 | ||
BY720838.1 | - | 3 | 487 |