Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpsm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029522.2 | NP_083798.2 | 13 | 1989 | ||
GenBank mRNA | AK018084.1 | - | 3 | 236 | ||
AK040996.1 | - | 13 | 1878 | |||
AK143552.1 | - | 13 | 1990 | |||
AY081187.1 | - | 13 | 1833 | |||
BC021308.1 | - | 12 | 1544 | 5007832 |