siRNA Details
Gene Symbol: Bcl7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029850.3 | NP_084126.1 | 3 | 386 | ||
GenBank mRNA | AK014498.1 | - | 3 | 329 | ||
AK044782.1 | - | 3 | 357 | |||
AK086803.1 | - | 3 | 383 | |||
AK157720.1 | - | 3 | 335 | |||
BC017640.1 | - | 3 | 300 | 4482250 | ||
BC038481.1 | - | 3 | 311 | 4501205 | ||
BC046777.1 | - | 3 | 216 | 5709511 | ||
BY271432.1 | - | 3 | 386 |

More Information
Other Species Targeted

