Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029654.4 | NP_083930.5 | 41 | 6329 | ||
GenBank mRNA | AK010577.1 | - | 4 | 493 | ||
AK220468.1 | - | 4 | 4956 | |||
BC024533.1 | - | 9 | 951 | |||
BC046427.1 | - | 20 | 2440 | |||
BC062182.1 | - | 41 | 6299 | 6505402 | ||
CA751117.1 | - | 4 | 374 |