Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps33a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029929.3 | NP_084205.3 | 2, 3 | 210 | ||
GenBank mRNA | AF439858.1 | - | 2, 3 | 194 | ||
AK019463.1 | - | 2, 3 | 194 | |||
AK041982.1 | - | 2, 3 | 206 | |||
AK140553.1 | - | 2, 3 | 199 | |||
AK150282.1 | - | 2, 3 | 202 | |||
AK150550.1 | - | 2, 3 | 202 | |||
AK194964.1 | - | 2, 3 | 124 | |||
BB852750.1 | - | 2, 3 | 234 | |||
BC046417.1 | - | 2, 3 | 201 | 4501207 |