Search
Search

Trim35| Gene Symbol: Trim35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029979.3 | NP_084255.2 | 1 | 512 | ||
| GenBank mRNA | AB060155.1 | - | 1 | 456 | ||
| AF145374.1 | - | 1 | 496 | |||
| AK003000.1 | - | 1 | 471 | |||
| AK020775.1 | - | 1 | 473 | |||
| AK031803.1 | - | 1 | 466 | |||
| AK032635.1 | - | 1 | 485 | |||
| AK075877.1 | - | 1 | 483 | |||
| AK129288.1 | - | 1 | 457 | |||
| AK163749.1 | - | 1 | 448 | |||
| BC049105.1 | - | 1 | 462 | 6418046 | ||
| BC053494.1 | - | 1 | 377 | |||
| BC145783.1 | - | 1 | 506 | 40131086 | ||
| BC145809.1 | - | 1 | 506 | 40131112 | ||