Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spink12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030061.3 | NP_084337.1 | 2, 3 | 272 | ||
GenBank mRNA | AK020352.1 | - | 2, 3 | 173 | ||
AK136616.1 | - | 2, 3 | 273 | |||
BC145914.1 | - | 2, 3 | 158 | 40131217 | ||
BC145916.1 | - | 2, 3 | 158 | 40131219 | ||
DQ437332.1 | - | 2, 3 | 331 |