Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031389.2 | NP_113566.2 | 3 | 1001 | ||
GenBank mRNA | AF285581.1 | - | 1 | 269 | ||
AK133284.1 | - | 3 | 1003 | |||
AK166817.1 | - | 3 | 982 | |||
AK166869.1 | - | 3 | 1227 | |||
AY596194.1 | - | 3 | 1001 | |||
BC120833.1 | - | 1 | 262 | 40129756 |