Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031392.2 | NP_113569.1 | 2 | 328 | ||
GenBank mRNA | AB041854.1 | - | 2 | 397 | ||
AF348591.1 | - | 2 | 324 | |||
AK037181.1 | - | 2 | 305 | |||
AK046119.1 | - | 2 | 295 | |||
AK052235.1 | - | 2 | 267 | |||
AK146617.1 | - | 2 | 285 | |||
AK163752.1 | - | 2 | 261 | |||
AK166265.1 | - | 2 | 305 | |||
AK169357.1 | - | 2 | 329 | |||
BC050894.1 | - | 2 | 328 | 5708880 | ||
BC054367.1 | - | 2 | 256 | 5346943 |