Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000328.2 | NP_000319.1 | 16 | 2105 | ||
XM_005272633.2 | XP_005272690.1 | 14 | 1801 | |||
XM_011543940.2 | XP_011542242.1 | 16 | 2101 | |||
XM_017029710.1 | XP_016885199.1 | 15 | 2749 | |||
XM_017029711.1 | XP_016885200.1 | 15 | 2746 | |||
XM_017029712.1 | XP_016885201.1 | 14 | 1798 | |||
GenBank mRNA | AJ238395.1 | - | 17 | 2031 | ||
AK223491.1 | - | 16 | 2079 | |||
AK291832.1 | - | 16 | 2112 | |||
BC031624.1 | - | 13 | 1651 | |||
U57629.1 | - | 16 | 1996 | |||
X97668.1 | - | 16 | 1937 |