siRNA Details
Gene Symbol: Pard3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122850.1 | NP_001116322.1 | 1, 2 | 426 | ||
NM_033620.2 | NP_296369.2 | 1, 2 | 426 | |||
GenBank mRNA | AK052416.1 | - | 1, 2 | 413 | ||
AK053563.1 | - | 1, 2 | 407 | |||
AK053576.1 | - | 1, 2 | 384 | |||
AK053762.1 | - | 1, 2 | 413 | |||
AK137690.1 | - | 1, 2 | 385 | |||
AY026057.1 | - | 1, 2 | 237 | |||
BC090616.1 | - | 1, 2 | 426 | 30943099 | ||
BC141187.1 | - | 1, 2 | 188 | 9055799 | ||
BC145508.1 | - | 1, 2 | 188 | 9054071 |

More Information
Other Species Targeted

