Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053129.3 | NP_444359.3 | 1 | 436 | ||
GenBank mRNA | AK029277.1 | - | 1 | 428 | ||
AK036169.1 | - | 1 | 437 | |||
AK143280.1 | - | 1 | 437 | |||
AK160730.1 | - | 1 | 437 | |||
AY013786.1 | - | 1 | 252 | |||
BC053098.1 | - | 1 | 409 | 6401988 |